Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810107E

Avanti

NBD AEA

Avanti Research - A Croda Brand 810107E, ethanol solution

Synonym(s):

20-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino] arachidonoyl ethanolamide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H39 N5O5
CAS Number:
Molecular Weight:
525.64
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>95% (TLC)

form

ethanol solution

packaging

pkg of 1 × 1 mL (810107E-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810107E

concentration

1 mg/mL (810107E-1mg)

shipped in

dry ice

storage temp.

−70°C

General description

The endocannabinoid pathway influences behavior, lipogenesis, embryologic development, and pathophysiological effects including pain, inflammation, stress, obesity, and non-alcoholic fatty liver disease. Endogenous endocannabinoids including arachidonoylethanolamide (anandamide, AEA) and 2-arachidonoylglycerol (2-AG) are derived from arachidonic acid (AA). Fluorescent NBD-AA and NBD-AEA analogs are direct probes that can be used for visualizing the uptake, intracellular trafficking, and targeting of these molecules by living cells, as well as measuring the binding of endocannabinoids to cytosolic binding proteins. Importantly, direct binding studies show the affinities of the fluorescent analogs are in the same range as the respective unlabeled ligands. Fluorescent endocannabinoid probes are useful for mapping the impact of various treatments, inhibitors, etc on endocannabinoid uptake and distribution in living cells.

Application

NBD AEA or 20-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino] arachidonoyl ethanolamide might be used:
  • to covalently label nanoparticles for fluorescence imaging experiments[1]
  • to develop probes for the determination of direct binding affinities for wild-type (WT) human fatty acid binding protein 1 (FABP1 T94T) or FABP1 T94A analogous to that for NBD-stearic acid binding to WT rat FABP1.4[2]
  • in fluorescent ligand displacement assays to confirm the binding of phytocannabinoids and synthetic cannabinoids to FABP1[3]

Packaging

5 mL Amber Glass Screw Cap Vial with Foil Bag Argon overseal (810107E-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

55.4 °F - closed cup

flash_point_c

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friedhelm Schroeder et al.
Lipids, 51(6), 655-676 (2016-04-28)
The first discovered member of the mammalian FABP family, liver fatty acid binding protein (FABP1, L-FABP), occurs at high cytosolic concentration in liver, intestine, and in the case of humans also in kidney. While the rat FABP1 is well studied
Gregory G Martin et al.
Biochemistry, 56(38), 5147-5159 (2017-08-31)
Using recombinant human wild-type fatty acid binding protein 1 (WT FABP1 T94T) and a variant (FABP1 T94A) protein, fluorescence binding assays, and circular dichroism, it was shown for the first time that WT FABP1 and the T94A variant each have
Christian Kleusch et al.
Molecules (Basel, Switzerland), 17(1), 1055-1073 (2012-01-24)
In this paper a rapid and highly efficient method for controlled incorporation of fluorescent lipids into living mammalian cells is introduced. Here, the fluorescent molecules have two consecutive functions: First, they trigger rapid membrane fusion between cellular plasma membranes and
Andrew Draganski et al.
The journal of sexual medicine, 15(5), 645-653 (2018-04-28)
Curcumin, a naturally occurring anti-inflammatory compound, has shown promise in pre-clinical studies to treat erectile dysfunction (ED) associated with type-1 diabetes. However, poor bioavailability following oral administration limits its efficacy. The present study evaluated the potential of topical application of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service