Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

800731P

Avanti

N-18:1 L-Serine MeEster

N-oleoyl L-serine methyl ester, powder

Synonym(s):

1-(9Z-octadecenoyl)-L-serine methyl ester; (S)-methyl-3-hydroxy-2-(9Z)-octadecenamidopropanoate; (S)-methyl-3-hydroxy-2-oleamidopropanoate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H41NO4
CAS Number:
Molecular Weight:
383.57
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (800731P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800731P

lipid type

phosphoglycerides
coenzymes

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (800731P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service