Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

792523C

Avanti

18:1(n9)- 16:0 PS - IsoPure

Avanti Research - A Croda Brand

Synonym(s):

1-oleoyl-2-palmitoyl-sn-glycero-3-phospho-L-serine (sodium salt, IsoPure)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H75NNaO10P
CAS Number:
Molecular Weight:
783.99
UNSPSC Code:
51191904
NACRES:
NA.25

assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

5 mg/mL in chloroform

technique(s)

mass spectrometry (MS): suitable

application(s)

lipidomics
metabolomics

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C40H76NO10P.Na/c1-3-5-7-9-11-13-15-17-18-20-21-23-25-27-29-31-38(42)48-33-36(34-49-52(46,47)50-35-37(41)40(44)45)51-39(43)32-30-28-26-24-22-19-16-14-12-10-8-6-4-2;/h17-18,36-37H,3-16,19-35,41H2,1-2H3,(H,44,45)(H,46,47);/q;+1/p-1/b18-17-;/t36-,37+;/m1./

InChI key

QKPXXPNOIWGYEJ-JEAFVVATSA-M

Packaging

5 mL Clear Glass Sealed Ampule (792523C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service