Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

791225P

Avanti

14:0 PE-DTPA(Cu)

Avanti Research - A Croda Brand 791225P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (copper salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C47H94CuN7O17P
CAS Number:
Molecular Weight:
1123.81
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (791225P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791225P

shipped in

dry ice

storage temp.

−20°C

General description

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid or 14:0 PE-DTPA is a synthetic phosphatidylethanolamine derivative.[1]
Inclusion of copper (Cu2+) ions in bicelles in the form of a lipid chelate resulted in a 10-fold T1 reduction and 2.7-fold increase in S/N of spectra for static experiments. This improvement would allow 2D experiments on membrane proteins in aligned lipid bilayers that typically take 3 days to be completed in <1.5 days. The technique is particularly beneficial to the study of membrane disrupting/permeating systems (such as antimicrobial peptides, toxins, amyloid peptides, and fusion peptides) whose concentration cannot be increased, as they result in nonlamellar conditions.

Application

14:0 PE-DTPA(Cu) may be usedis suitable for use for liposome preparations[2] and liposome labelling.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (791225P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recognition of lipopeptide patterns by Toll-like receptor 2-Toll-like receptor 6 heterodimer
Kang JY, et al.
Immunity, 31(6), 873-884 (2009)
Kazutoshi Yamamoto et al.
Journal of the American Chemical Society, 132(20), 6929-6931 (2010-05-04)
Recent studies have demonstrated the abilities of solid-state NMR techniques to solve atomic-level-resolution structures and dynamics of membrane-associated proteins and peptides. However, high-throughput applications of solid-state NMR spectroscopy are hampered by long acquisition times due to the low sensitivity of
Direct observation of the three regions in $\alpha$-synuclein that determine its membrane-bound behaviour
Fusco Giuliana, et al.
Nature Communications, 5, 3827-3827 (2014)
Tumor targeting and imaging with dual-peptide conjugated multifunctional liposomal nanoparticles
Rangger C, et al.
International journal of nanomedicine, 8(6), 4659-4659 (2013)
Rupa R Sawant et al.
Pharmaceutical research, 29(2), 375-383 (2011-08-17)
To evaluate and compare anticancer therapeutic effect of palmitoyl ascorbate liposomes (PAL) and free ascorbic acid (AA). Liposomes incorporating palmitoyl ascorbate (PA) were prepared and evaluated for PA content by HPLC. To elucidate mechanism of action of cell death in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service