Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

790473P

Avanti

06:0 PC-d35

Avanti Research - A Croda Brand 790473P, powder

Synonym(s):

1,2-dihexanoyl-d22-sn-glycero-3-phosphocholine-1,1,2,2-d4-N,N,N-trimethyl-d9

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H5NO8PD35
CAS Number:
Molecular Weight:
488.72
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (790473P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790473P

lipid type

deuterated lipids
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Packaging

5 mL Amber Glass Screw Cap Vial (790473P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominique Mias-Lucquin et al.
Journal of structural biology, 209(1), 107411-107411 (2019-11-07)
Dystrophin is a large intracellular protein that prevents sarcolemmal ruptures by providing a mechanical link between the intracellular actin cytoskeleton and the transmembrane dystroglycan complex. Dystrophin deficiency leads to the severe muscle wasting disease Duchenne Muscular Dystrophy and the milder

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service