Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700319P

Avanti

Chenodeoxycholic acid disulfate trisodium salt

Avanti Research - A Croda Brand

Synonym(s):

Chenodeoxycholic acid disulfate trisodium salt, 3α, 7α -dihydroxy-5β-cholan-24-oic acid disulfate trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H37Na3O10S2
Molecular Weight:
618.64
UNSPSC Code:
12161700
NACRES:
NA.25
Product 700319P is not currently sold in your country. Contact Technical Service

assay

>99% (TLC)

Quality Level

form

powder

packaging

package of 1 × 5 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bile acids

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (700319P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service