700294O
Avanti
Farnesol-d6
Avanti Research™ - A Croda Brand
Synonym(s):
(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol(d6)
Select a Size
Select a Size
About This Item
Recommended Products
form
oil
packaging
package of 1 × 1 mg
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C
SMILES string
C/C(CC/C=C(C)/CCC=C(C([2H])([2H])[2H])C([2H])([2H])[2H])=C\CO
InChI key
CRDAMVZIKSXKFV-JPWGLETFSA-N
Packaging
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service