Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

700225P

Avanti

3β-Hydroxy-7-oxocholest-5-enoic acid-d3

Avanti Research - A Croda Brand

Synonym(s):

Cholest-5-en-26-oic-27,27,27-d3 acid, 3-hydroxy-7-oxo-, (3β)-

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H39D3O4
CAS Number:
Molecular Weight:
433.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

assay

99% (TLC)

form

powder

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

3β-Hydroxy-7-oxocholest-5-enoic acid-d3 is a deuterated derivative of 3β-Hydroxy-7-oxocholest-5-enoic acid.

Packaging

5 mL Amber Glass Screw Cap Vial (700225P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service