Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

700182M

Avanti

7αOH (D7) Chol ester

methylene chloride solution

Synonym(s):

25,26,26,26,27,27,27-heptadeuteriocholest-5-ene-3β,7a-diol (9Z-octadecenoate)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C45H71D7O3
Molecular Weight:
674.14
MDL number:
UNSPSC Code:
12352211

assay

99% (TLC)

form

methylene chloride solution

packaging

package of 1 × 1 mg (700182M-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL in methylene chloride

shipped in

dry ice

storage temp.

−20°C

General description

25,26,26,26,27,27,27-heptadeuteriocholest-5-ene-3β,7a-diol (9Z-octadecenoate) or 7αOH (D7) Chol ester is a deuterated derivative of 7αOH Chol ester.

Packaging

5 mL Clear Glass Sealed Ampule (700182M-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service