Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700130P

Avanti

3β,24S-dihydroxy-5-cholestenoic acid

Avanti Research - A Croda Brand 700130P, powder

Synonym(s):

(25R)-cholest-5-en-26-oic acid,3β,24S-dihydroxy

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H44O4
CAS Number:
Molecular Weight:
432.64
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700130P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700130P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H](C1)CC[C@](C1=CC2)(C)C3C2C(CC[C@@H]4[C@H](C)CC[C@H](O)[C@@H](C)C(O)=O)[C@]4(C)CC3

InChI key

XXUCPCOTPNGCMA-ODGUSDSXSA-N

General description

3β,24S-dihydroxy-5-cholestenoic acid is a cholestenoic acid (CA). The enzyme sterol 27-hydroxylase (CYP27A1) catalyzes the synthesis of CA in the mitochondria.[1]

Biochem/physiol Actions

Cholestenoic acid (CA) is a ligand to nuclear receptor. Based on the structural features, CA may elicit neurotoxic or neuroprotective effects.[2] Defects in the enzymes catalyzing the formation of cholestenoic acids are implicated in disorders related to motor neuron degeneration.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (700130P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholestenoic acids regulate motor neuron survival via liver X receptors
Theofilopoulos S, et al
The Journal of Clinical Investigation, 124, 4829-4842 (2014)
Formation and metabolism of oxysterols and cholestenoic acids found in the mouse circulation: Lessons learnt from deuterium-enrichment experiments and the CYP46A1 transgenic mouse
Crick PJ, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 195, 105475-105475 (2019)
Cholestenoic acid is a prognostic biomarker in acute respiratory distress syndrome
Madenspacher JH, et al.
The Journal of Allergy and Clinical Immunology, 143, 440-442 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service