Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

700091P

Avanti

dihydrolanosterol-d7

Avanti Research - A Croda Brand

Synonym(s):

111017

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H45D7O
CAS Number:
Molecular Weight:
435.78
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.12

description

25,26,26,26,27,27,27-heptadeuteriolanost-8-en-3β-ol

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700091P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Dihydrolanosterol is a lanolin component present along with lanosterol and cholesterol.[1] It constitutes as 40–50% impurity in commercial lanosterol.[2] Dihydrolanosterol-D7 is the deuterated derivative of dihydrolanosterol.

Packaging

5 mL Amber Glass Screw Cap Vial (700091P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levan K Kavtaradze et al.
Steroids, 69(10), 697-700 (2004-10-07)
We describe an inexpensive, low-toxicity and high-yielding method for the production of pure lanosterol and dihydrolanosterol from the commercially available mixture. Optimum conditions are presented for the one-pot production of the intermediate 24,25 vicinal diol of lanosterol acetate (via either
Hairun Pei et al.
Journal of separation science, 42(12), 2171-2178 (2019-04-06)
Lanosterol is a potential drug for cataracts treatment, which can reverse the aggregation of intracrystalline proteins. The low concentration in lanolin calls for high-performance separation methods. In this study, a counter-current chromatography dual-mode elution method was developed for the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service