Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700084P

Avanti

27-hydroxy cholestenone

Avanti Research - A Croda Brand

Synonym(s):

(25R)-cholest-4-ene, 26-hydroxy-3-oxo

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
UNSPSC Code:
12352211
NACRES:
NA.25

description

27-hydroxy-4-cholesten-3-one

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700084P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

Storage temp.

−20°C

SMILES string

CC1(CC2)C(CCC3C1CCC4(C)C3CCC4C(CCCC(C)CO)C)=CC2=O

General description

Cholestenone differs from cholesterol by having a keto group in the place of 3-hydroxyl group.It is a poorly catabolized metabolite.[1] 27-hydroxy cholestenone is synthesized from cholesterol and this oxidation step is catabolized by the enzyme cholesterol oxidase.[2]

Biochem/physiol Actions

Cholestenone is oxidized by bacteria as a nutritional source. Liver is the site of cholestenone metabolism in mammals. [1]

Packaging

5 mL Amber Glass Screw Cap Vial (700084P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Norlin et al.
Journal of lipid research, 44(8), 1515-1522 (2003-06-05)
The mitochondrial sterol 27-hydroxylase (CYP27A1) is required for degradation of the C27-sterol side chain in bile acid biosynthesis. CYP27A1 seems, however, to have roles beyond this, as illustrated by patients with a deficient sterol 27-hydroxylase due to mutations of the
Maarit Neuvonen et al.
PloS one, 9(8), e103743-e103743 (2014-08-27)
Bacterial cholesterol oxidase is commonly used as an experimental tool to reduce cellular cholesterol content. That the treatment also generates the poorly degradable metabolite 4-cholesten-3-one (cholestenone) has received less attention. Here, we investigated the membrane partitioning of cholestenone using simulations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service