Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700012P

Avanti

15α-hydroxycholestane

Avanti Research - A Croda Brand

Synonym(s):

3b,15a-diol-5a-cholestane

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H48O2
CAS Number:
Molecular Weight:
404.67
UNSPSC Code:
12352211

description

5α-cholestane-3β,15α-diol

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700012P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

15α-hydroxycholestane is a derivative of cholesterol and is used as a standard in Liquid chromatography-mass spectrometry (LC-MS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700012P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comprehensive method for extraction and quantitative analysis of sterols and secosteroids from human plasma
McDonald JG, et al.
Journal of Lipid Research, 53(7), 1399-1409 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service