699200P
Avanti
Lipid A - Purified
Avanti Research™ - A Croda Brand
Synonym(s):
Lipid A Detoxified (Salmonella minnesotaR595)
Select a Size
Select a Size
About This Item
Recommended Products
assay
>99% (HPLC)
form
powder
packaging
pkg of 1 × 1 mg (699200P-1mg)
pkg of 1 × 5 mg (699200P-5mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C
SMILES string
O[C@H]([C@@H](CO[C@@H]([C@@H]1NC(C[C@H](OC(CCCCCCCCCCC)=O)CCCCCCCCCCC)=O)O[C@H](CO)[C@@H](OP([O-])(O)=O)[C@@H]1OC(C[C@H](O)CCCCCCCCCCC)=O)O[C@H](O)[C@@H]2NC(C[C@H](O)CCCCCCCCCCC)=O)[C@@H]2OC(C[C@H](O)CCCCCCCCCCC)=O.[NH4+]
General description
Application
Biochem/physiol Actions
Packaging
Other Notes
Legal Information
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service