Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

640011P

Avanti

Ceramide Synthase (CerS1/CerS4) Assay Kit

Avanti Research - A Croda Brand

Synonym(s):

Ceramide Synthase (CerS1/CerS4) Assay Kit, CerS1/S4 Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12161503

form

powder

usage

sufficient for ~275 reactions

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

The CerS Assay Kit provides an easy to use assay for measuring CerS activity in biological samples. This Kit offers the advantage of a short assay time requiring only small amounts of biological material while eliminating TLC as a separation technique with its attendant degradation of NBD sphinganine.

Packaging

Kit contains:
0.3mg 18:0 fatty acyl-CoA
0.1mg NBD Sphinganine

Storage and Stability

Note: this product should be stored at -20° for up to 6 months.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 810206PNBD Sphinganine, omega(7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R)-2-aminooctadecane-1,3-diol, powderSDS

  • 870718P18:0 Coenzyme A, Avanti Research - A Croda Brand 870718P, powderSDS

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service