Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

600510

Avanti

5mL Amber SCHOTT® FIOLAX® Vial with PTFE-lined Screw Cap Closure

Avanti Research - A Croda Brand

Synonym(s):

SCHOTT® FIOLAX® Vial

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352211

packaging

pkg of 1 vial

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

General description

Lipid storage product.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
FIOLAX is a registered trademark of Schott AG
Schott is a registered trademark of Schott AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service