Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

600320AVT

Avanti

125 mL Boston Round

Avanti Research - A Croda Brand

Synonym(s):

120 mL amber narrow mouth bottle with PTFE lined screw cap closure

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
24122000
NACRES:
NA.25

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

Packaging

pkg of 1 x 120 mL bottle (600320-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service