550002P
Avanti
Aurora™-PLC
Avanti Research™ - A Croda Brand 550002P, powder
Synonym(s):
Aurora™-PLC, Aurora™-PLC Nanoparticles
About This Item
Recommended Products
assay
>95% (TLC)
form
powder
packaging
pkg of 1 × 10 mg (with screw cap (550002P-10mg))
pkg of 1 × 100 mg (with screw cap (550002P-100mg))
manufacturer/tradename
Avanti Research™ - A Croda Brand 550002P
shipped in
dry ice
storage temp.
−20°C
General description
Packaging
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service