Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

360500

Avanti

Huzzah KLA

Avanti Research - A Croda Brand

Synonym(s):

Huzzah KLA, Human serum albumin / Kdo2 Lipid A complex

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 vial (360500-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Avanti′s new HSA lipid delivery system Huzzah, enhances the cellular delivery of lipid species by conjugating with human serum albumin (HSA). HSA is the most applicable carrier protein.[1] This conjugate system eliminates the need for organic solvents, which are well-known to adversely affect viability and phenotypic characters of cells. Avanti′s latest conjugate, Huzzah KLA, provides the researcher with the ability to deliver this new synthetic glycan in aqueous solution. Many lipid A- based adjuvant systems require lengthy sonication and serial dilution. Huzzah KLA is provided as a freeze-dried conjugate with HSA that requires only the addition of saline to be ready for use.

Packaging

10 mL Clear Glass Vial with Stopper and Crimp Cap (360500-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Huzzah is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avanti lipid tools: connecting lipids, technology, and cell biology
Sims KH, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1841(8), 1038-1048 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service