360493P
Avanti
Huzzah™ C1P
Avanti Research™ - A Croda Brand
Synonym(s):
Huzzah™ C1P, HUMAN SERUM ALBUMIN / CERAMIDE 1 PHOSPHATE COMPLEX
Select a Size
Select a Size
About This Item
Recommended Products
form
powder
packaging
pkg of 1 × 1 ea (360493P-1EA)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C
General description
*Exact concentration will vary from batch to batch. Refer to label for exact product concentration upon reconstitution as determined by mass spectrometric analysis.
Molar Ratio: C1P:albumin (1:1) Albumin Biological Characterization: microbial free; endotoxin < 1 IU/mL Albumin Lipid Characterization: ~0.07g/L phospholipids; ~0.02 g/L sphingolipids; ~0.1 g/L fatty acids; and triglycerides and cholesterols below the limit of quantitation of 0.1 g/L and 0.05 g/L
Packaging
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service