360492P
Avanti
Huzzah™ S1P
Avanti Research™ - A Croda Brand
Synonym(s):
Huzzah™ S1P, Human Serum Albumin/Sphingosine-1-Phosphate Complex
Select a Size
Select a Size
About This Item
Recommended Products
assay
>99% (MS)
form
powder
manufacturer/tradename
Avanti Research™ - A Croda Brand
packaging
1 ea of 1 × (with stopper and crimp cap)
shipped in
dry ice
Storage temp.
−20°C
General description
Application
Packaging
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service