Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330711X

Avanti

Odd-Chained LIPIDOMIX Mass Spec Standard

Avanti Research - A Croda Brand

Synonym(s):

LIPIDOMIX Mass Spec Standard, solution (1:1 dichloromethane:methanol)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41116107

form

solution (1:1 dichloromethane:methanol)

packaging

ampule of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

2 mL amber glass sealed ampule

Components

The Odd-Chained LIPIDOMIX Mass Spec Standard is supplied in 1 mL of (1:1) dichloromethane:methanol and contains the following components:

Mixture ComponentTarget Conc. (μM)
  • 17:1 Lyso PG(Na salt)25
  • 17:1 Lyso PA(NH4 salt)36
  • 17:1 Lyso PI (NH4 salt)22
  • 17:1 Lyso PS(Na salt)25
  • 17:1 Lyso PC1125
  • 17:1 Lyso PE25
  • 17:0-17:0 DAG500
  • 17:0-17:0-17:0 TAG1750
  • 12:0 SM (d18:1/12:0)1000
  • 17:0-14:1 PC5250
  • 17:0-14:1 PS (NH4 salt)250
  • 17:0-14:1 PG (NH4 salt)125
  • 17:0-14:1 PA (NH4 salt)25
  • 17:0-14:1 PE 175
  • 17:0-14:1 PI (NH4 salt)250
  • 17:0 cholesterol ester13250

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

target_organs

Central nervous system, Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service