Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330503

Avanti

Sphingolipid Snoopers

Avanti Research - A Croda Brand

Synonym(s):

Sphingolipid Snoopers

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352211

description

Sphingolipid Snoopers

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 5 × (membrane packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti Sphingolipid Snoopers consist of 14 different lipid species spotted individually on a solid support. Each spot contains 1 μg of the highest quality pure lipid and is ideal for investigating lipid-protein interactions.

application

Sphingolipid Snoopers has been used as a positive control for the detection of lipid peroxidation.[1]

Packaging

Membrane Packaged in Foil Pouch (330503-5EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noemi Jiménez-Rojo et al.
Biochimica et biophysica acta, 1838(8), 2071-2077 (2014-05-08)
Lipid peroxidation plays a central role in the pathogenesis of many diseases like atherosclerosis and multiple sclerosis. We have analyzed the interaction of sphingosine with peroxidized bilayers in model membranes. Cu(2+) induced peroxidation was checked following UV absorbance at 245nm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service