Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330502

Avanti

BMP Snoopers

Avanti Research - A Croda Brand

Synonym(s):

BMP Snoopers, Bis(Monoacylglycerol) Phosphate Snoopers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.62

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 5 × (membrane packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti BMP Snoopers consist of 9 different lipid species spotted individually on a solid support. Each spot contains 1 μg of the highest quality pure lipid and is ideal for investigating lipid-protein interactions.

Packaging

Membrane Packaged in Foil Pouch (330502-5EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service