Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330501

Avanti

Oxidized Phospholipid Snoopers

Oxidized Phospholipid Snoopers

Synonym(s):

Oxidized Phospholipid Snoopers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.62

manufacturer/tradename

Avanti Research - A Croda Brand 330501

packaging

1 ea of 5 × (membrane packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti Ox-Phospholipid Snoopers consist of 12 different lipid species spotted individually on a solid support. Each spot contains 1 μg of the highest quality pure lipid and is ideal for investigating lipid-protein interactions.

Application

Oxidized Phospholipid Snoopers has been used in in vitro biorthogonal reaction for analyzing the specificity of antibody- trans-cyclooctene (Ab-TCO).
Oxidized Phospholipid Snoopers has been used to study lipid-protein interactions, characterize lipid-binding antibodies and identify novel lipid-interacting proteins of potential therapeutic value.

Packaging

Membrane Packaged in Foil Pouch (330501-5EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unambiguous detection of atherosclerosis using bioorthogonal nanomaterials
Pellico J, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 17, 26-35 (2019)
Oriol Gallego et al.
Molecular systems biology, 6, 430-430 (2010-12-02)
Protein-metabolite networks are central to biological systems, but are incompletely understood. Here, we report a screen to catalog protein-lipid interactions in yeast. We used arrays of 56 metabolites to measure lipid-binding fingerprints of 172 proteins, including 91 with predicted lipid-binding
Ronald C Bruntz et al.
The Journal of biological chemistry, 289(2), 600-616 (2013-11-22)
The lack of innovative drug targets for glioblastoma multiforme (GBM) limits patient survival to approximately 1 year following diagnosis. The pro-survival kinase Akt provides an ideal target for the treatment of GBM as Akt signaling is frequently activated in this
Avanti lipid tools: connecting lipids, technology, and cell biology
Sims KH, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1841(8), 1038-1048 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service