Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330006S

Avanti

MB47

Avanti Research - A Croda Brand

Synonym(s):

MB47 Mouse Monoclonal Antibody (IgG) Anti-(Human Apo-B100), solution

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.25

antibody product type

primary antibodies

form

solution

packaging

pkg of 1 × 100 μg (330006S-100UG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL

shipped in

dry ice

storage temp.

−70°C

General description

Apolipoproteins B (apoB) is a large (4538 amino acids), amphipathic glycoprotein. It is encoded by the gene APOB, mapped to human chromosome 2. A unique mRNA editing process results in the formation of two structurally related by distinct isoforms of apo-B, includes apoB-48 and apoB-100. ApoB-100 is synthesized in the liver. apoB has both hydrophobic lipid binding regions and hydrophilic region that interacts with plasma. It is arranged as α-helices and β-sheet domains, designated NH2-βα1-β1-α2-β2-α3-COOH. The mature protein has 25 cysteine residues, out of which 16 residues are involved in intramolecular disulfide bridges and proper folding of the amino terminal end of the protein.[1] MB47 binds near to 3500 amino acid of apoB-100.[2]

Biochem/physiol Actions

Apolipoproteins B (apoB) plays a key role in mammalian lipoprotein metabolism. ApoB-100 is the structural protein involved in triglyceride rich very low sdensity lipoprotein (VLDL) synthesis. apo-B100 is the protein in cholesteryl ester-enriched low-density lipoprotein (LDL). High plasma levels of apo-B100 is involved in premature atherosclerosis.[3] apo-B100 functions as a ligand for receptor mediated endocytosis of LDL.[1]

Packaging

0.5 mL Polypropylene Tube with Screw Cap (330006S-100UG)

Physical form

1X PBS (Phosphate Buffered Saline) with 0.27 mM Na2 EDTA

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimyristoylphosphotidylcholine induces conformational changes in apoB that lowers lipoprotein (a)
Wang YT, et al.
Journal of Lipid Research, 50(5), 846-853 (2009)
Transgenic mice expressing high plasma concentrations of human apolipoprotein B100 and lipoprotein (a).
Linton MF, et al.
The Journal of Clinical Investigation, 92(6), 3029-3037 (1993)
Lipid disorders and mutations in the APOB gene
Whitfield AJ, et al.
Clinical Chemistry, 50(10), 1725-1732 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service