330003S
Avanti
E06 mAb-TopFluor™
E06 Mouse Monoclonal Antibody (IgM), TopFluor™ labeled Anti-(Oxidized Phospholipid), 1X TBS (Tris Buffered Saline) with 1%BSA
Synonym(s):
E06 mAb-TopFluor™, T15 antibody
Select a Size
Select a Size
About This Item
Recommended Products
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
assay
>95%
form
solution
packaging
pkg of 1 × 0.1 mL (polypropylene tube with screw cap (330003S-25ug))
manufacturer/tradename
Avanti Research™ - A Croda Brand 330003S
concentration
0.25 mg/mL (330003S-25ug)
shipped in
dry ice
storage temp.
−70°C
General description
Application
- in the detection of oxidized phospholipids by immunocytochemical method in human primary skin fibroblasts[2]
- as a component of blocking solution in competitive binding assay of oxidized phosphatidylcholine[1]
- for the detection of oxidized phospholipids (OxPL) in the extracellular vesicle by flow cytometry[3]
Biochem/physiol Actions
Packaging
Preparation Note
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service