Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300106S

Avanti

Control (5mL vial)

Avanti Research - A Croda Brand 300106S

Synonym(s):

Control, Control (Pegylated Liposomal Vehicle)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352202

form

solution

packaging

pkg of 1 × 5 mL (with stopper and crimp cap (300106S-1EA))

manufacturer/tradename

Avanti Research - A Croda Brand 300106S

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Particle size and shape: Vehicle control mean particle size is 80 to 90 nm, with very small poly-dispersity valueFormulation: The long-circulating, pegylated, liposomal carrier is composed of N-(carbonyl-ethoxypolyethylene glycol 2000)-1,2-distearoyl-sn-glycero-3-phosphoethanolamine sodium salt (MPEG-DSPE), 3.19 mg/ml; fully hydrogenated soy phosphatidylcholine (HSPC), 9.58 mg/ml; cholesterol, 3.19 mg/ml, ammonium sulfate, approximately 2 mg/ml; histidine as a buffer; hydrochloric acid or sodium hydroxide for pH control; and sucrose to maintain isotonicity. The Control is provided as a sterile, translucent, liposomal dispersion in single-use, 2 ml or 5 ml vials.Buffer: 10 mM histidine buffer, pH 6.5-6.8, with 10% w/v sucrose.

Application

Intravenous (i.v.) injection in flank or orthotopic animal models of cancer.

Packaging

10 mL Clear Glass Vial with Stopper and Crimp Cap (300106S-1EA)

Storage and Stability

Store undiluted at 2°-8° C. Do Not Freeze.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service