Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300103S

Avanti

Control (2mL vial)

Avanti Research - A Croda Brand 300103S

Synonym(s):

Control, Control (Pegylated Liposomal Vehicle)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

form

solution

packaging

pkg of 1 × 2 mL (with stopper and crimp cap (300103S-1EA))

manufacturer/tradename

Avanti Research - A Croda Brand 300103S

application(s)

advanced drug delivery

shipped in

wet ice

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Particle size and shape: Vehicle control mean particle size is 80 to 90 nm, with very small poly-dispersity valueFormulation: The long-circulating, pegylated, liposomal carrier is composed of N-(carbonyl-ethoxypolyethylene glycol 2000)-1,2-distearoyl-sn-glycero-3-phosphoethanolamine sodium salt (MPEG-DSPE), 3.19 mg/ml; fully hydrogenated soy phosphatidylcholine (HSPC), 9.58 mg/ml; cholesterol, 3.19 mg/ml, ammonium sulfate, approximately 2 mg/ml; histidine as a buffer; hydrochloric acid or sodium hydroxide for pH control; and sucrose to maintain isotonicity. The Control is provided as a sterile, translucent, liposomal dispersion in single-use, 2 ml or 5 ml vials.Buffer: 10 mM histidine buffer, pH 6.5-6.8, with 10% w/v sucrose.

Application

Intravenous (i.v.) injection in flank or orthotopic animal models of cancer.

Packaging

2 mL Clear Glass Vial with Stopper and Crimp Cap (300103S-1EA)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.
Store undiluted at 2°-8° C. Do Not Freeze.

Other Notes

Delivery to cells: Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration: Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service