Key Documents
Z804401
Sigma-Aldrich
E-Z Quick Table Top KBr Pellet Press w/2mm, 4mm and 7mm die set
About This Item
UNSPSC Code:
23151817
NACRES:
SB.32
technique(s)
IR spectroscopy: suitable
General description
The Quick Table Top Press operates on the same toggle action principle as our Quick Press (Z506761). By mounting the press vertically and providing a table top mounting platform, it is now easier to compress a KBr pellet and to open the press and remove the pellet. Vertical mounting enables the use of body weight and shoulder muscles, so the press requires minimal strength to operate. When the base is attached to a bench it is easier to open then a hand press, and pushing a small lever ejects the die set. There is a dial indicator to enable reproduction of force settings from pellet to pellet. The press is constructed from Stainless Steel, chrome plated steel and anodized aluminum for long corrosion-free service.
Legal Information
E-Z is a trademark of International Crystal Laboratories Corp.
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)