Key Documents
Z804185
Sigma-Aldrich
Polystyrene calibration films for IR, NIST traceable
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
Product Z804185 is not currently sold in your country. Contact Technical Service
General description
Pair of polystyrene calibration films of different thickness. One film is 38 microns and is provided with certification of traceability to five (5) NIST 1921b frequencies. The second film is 76 microns and is provided without certification. Both films are mounted on 2" x 3" cardstock holder with a 19mm aperture for mounting in the universal slide holder in all instruments. Includes instructions for two (2) film ordinate calibration and storage case.
Choose from one of the most recent versions:
Certificates of Analysis (COA)