Key Documents
Z768448
ChillDown, for 250 mL beaker
pkg of 1 ea
About This Item
UNSPSC Code:
41121803
NACRES:
NB.35
material
polyethylene
packaging
pkg of 1 ea
manufacturer/tradename
Diversified Biotech CHDN-1000
capacity
250 mL
General description
Polyethylene beaker cooler filled with propylene glycol.
The ChillDown is ideal for rapidly cooling agarose solutions, chemical mixtures or any cooling application that can take place in a 250 mL beaker.
Unit should be charged by leaving it in a freezer for several hours (ideally, overnight).
Choose from one of the most recent versions:
Certificates of Analysis (COA)