Z767840
EUROSTAR 200 digital overhead stirrer
AC/DC input 230 V AC, UK plug
Synonym(s):
electronic overhead stirrers, laboratory mixers, laboratory stirrers
About This Item
Recommended Products
manufacturer/tradename
IKA 3990000
parameter
0/6-2000 rpm speed
W × H × D
91 mm × 274 mm × 209 mm
shaft diam. × L
16 mm × 220 mm
stirrer capacity
100 L
output
84 W
General description
- Digital speed display
- Infinitely adjustable speed
- Push-through agitator shafts
- Overload protection
- Short-term overload operation
- Slim casing
- Quiet operation
- Error code display
Choose from one of the most recent versions:
Certificates of Analysis (COA)