Key Documents
Z764604
Sigma-Aldrich
Stopcock Adapter
4 mm PTFE stopcock, bent, joint: ST/NS 34/45
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint: ST/NS34/45
General description
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)