Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z760870

Nichipet EX-Plus II pipette

volume 1000-5000 μL

Synonym(s):

Nichiryo pipette, liquid handling, pipettors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500

feature

electronic: no
single-channel

packaging

pkg of 1 ea

manufacturer/tradename

Nichiryo 00-NPL02-5000

volume

1000-5000 μL

Looking for similar products? Visit Product Comparison Guide

General description

The Nichipet EX-Plus ll is a new digital micropipette addition to the Nichiryo family of pipettors. Nichipet EX-Plus ll offers a new, key feature of one-touch calibration. Quick and easy.

Time and money spent on calibration of pipettes is now greatly reduced!

  • UV resistant
  • Solvent resistant
  • Fully autoclavable
  • One touch calibration
  • Ceramic pistons
  • Filtered nozzles
  • Easy, convenient locking lever
  • Ergonomic design
  • Single channel, manual, continuously adjustable volume

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service