Z760013
Ohaus® Navigator® balance, NV Series
Model NV511, AC/DC input 230 V AC (universal plugs)
Synonym(s):
analytical scales, laboratory balances, weighing balances
About This Item
Recommended Products
material
stainless steel plate
manufacturer/tradename
Ohaus® 83032292
weighing capacity measuring range
510 g, resolution: 0.1 g, linearity: ±0.2 g
parameter
20-85% humidity limit
W × depth × H
204 mm × 212 mm × 134 mm
platform W × L
190 mm × 138 mm
battery power
4 AA (batteries not included)
Looking for similar products? Visit Product Comparison Guide
General description
- Two touchless sensors free up your hands for handling samples and limiting keypad wear and tear.
- Ultra-fast stabilization in less than one second provides fast, accurate and repeatable weighing results, improving operator efficiency, productivity and throughput.
- Superior mechanical overload protection system & integrated shipping lock allows the scale to withstand loads up to 403% of its rated capacity, and protects the weighing cell from damage during transportation or storage.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service