Key Documents
Z758825
Sigma-Aldrich
ACD/Spectrus Processor with Aldrich® FT-NMR Library
pkg of 1 ea
Synonym(s):
nmr library
About This Item
UNSPSC Code:
41100000
packaging
pkg of 1 ea
manufacturer/tradename
ACD Labs
General description
ACD/Spectrus Processor enables access to all your analytical data and the Aldrich FT-NMR Library in one integrated software package With more than 35,000 spectra of real products available, chemists can quickly verify their starting materials, and search for unknown chemicals or impurities within the analytical data-processing interface.
Features
  • Perform peak and spectral similarity searches with your experimental NMR data
  • Search on exact structures or substructures through the ACD/ChemSketch interface
  • Search Aldrich related information such as CAS no., catalog number, boiling and melting points, density, refractive index, etc.
  • All search results are presented for direct comparison in the same processing window
Benefits
  • One software for all your analytical chemistry needs
  • Access all your analytical data (NMR, LC-MS, IR, etc.) in one processing interface
  • Faster data analysis and interpretation times
  • Check starting materials before beginning large/involved syntheses
  • Quickly identify unknowns and impurities
  • One software interface for data processing, database searching, and spectra comparison
Spectrus Processor allows more efficient management of the analytical data workflow, freeing chemists′ time for other important work. Contact us today for more information.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)