VACUU-LAN Local Vacuum Network technology can deliver vacuum down to 2 mbar (1.5 Torr) to bench vacuum ports. Vacuum is provided locally by a quiet, oil-free VACUUBRAND pump, such as the model MD 4C NT pumps, so there is no interference or risk of cross-contamination from other labs.
The system is completely modular; add vacuum one lab at a time, extend capability as needed, and even interchange vacuum ports. Individual ports are check-valve-controlled to minimize interference among applications. Ports include manual flow control for continuously variable pumping speed adjustment.
Higher flow-rate pumps can support two VACUU-LAN Mini-Networks at once, and pumps with deeper vacuum capacity, such as VACUUBRAND model PC 201 NT pumps, will assist with your evaporative applications.
Legal Information
VACUU•LAN is a registered trademark of Vacuubrand GmbH & Co. KG
Vacuubrand is a registered trademark of Vacuubrand GmbH & Co. KG