Z757136
Kern ionizer, neutralizes electrostatic charge, for DLB moisture analyzer
EU plug
Synonym(s):
air ionizers, ionizers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111502
description
Neutralization: +/- 20V
feature
Discharge of ozone: 0.04 ppm/h
EU plug
manufacturer/tradename
Kern YBI-01 EU
W × depth × H
97 mm × 53 mm × 31 mm
Looking for similar products? Visit Product Comparison Guide
General description
When weighing plastics, china, or glass, these materials often carry an electrostatic charge. The electromagnetic field that occurs between the material to be weighed and the balance can cause the weighing result to be distorted by up to a gram.
The Kern ionizer neutralizes the electrostatic charge providing a more accurate measurement in milligrams or even smaller increments.
The Kern ionizer neutralizes the electrostatic charge providing a more accurate measurement in milligrams or even smaller increments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service