Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z752088

ColiCheck with MUG

P/A test kit with sample bottles

Synonym(s):

10496745

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NB.79

packaging

pk of 30

manufacturer/tradename

Maine Mfg 10496745

application(s)

food and beverages

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service