Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z752061

Sigma-Aldrich

Whatman® Brilliant Green Bile Bottled Broth

with Durham tubes

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

packaging

pk of 20

manufacturer/tradename

Maine Mfg 10496710

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service