Key Documents
Z751987
FAST PAK protein array kits
1-pad 20 x 51 mm, 10 slides and components
About This Item
UNSPSC Code:
41105906
Pricing and availability is not currently available.
manufacturer/tradename
Whatman 10485262
Whatman Article No. 28416233 (US reference)
suitability
suitable for (ELISA format (sandwich assay) antibody arrays, protein arrays, carbohydrate arrays, reverse phase (micro-Western) arrays using cell or tissue lysates)
General description
FAST PAK protein array kits provide the necessary components for researchers to conveniently build and process protein microarrays.
FAST PAK kits are available for all FAST Slide formats (1, 2, 8, and 16 pads). Each kit contains 10 FAST Slides, 10 Incubation Chambers, 40 Chamber Covers, 10 ml 2 x Protein Arraying Buffer, 15 ml Protein Array Blocking Buffer, and 125 ml 10 x Protein Array Wash Buffer.
Note: A reusable slider holder is also needed, either FAST Frame for 1 to 4 slides or Chip Clip for 1 slide.
Features and benefits
  • 1, 2, 8, or 16 array pads on each slide
  • Each pad can be processed separately to increase the number of arrays on each slide and reduce sample volume
  • Suitable for multiplex experiments, side-by-side comparisons, and control experiments all on the same slide
  • Reproducible results from slide-to-slide and pad-to-pad
  • Increased protein stability and enhanced signal intensity with FAST PAK array buffer
  • Proprietary protein array blocking buffer ensures optimum signal-to-noise ratios and promotes specific binding
  • Flexibility for detection by fluorescent, chemiluminescent, colorimetric, and radiographic methods
Applications
  • ELISA format (sandwich assay) experiments using antibody arrays
  • Reverse phase (micro-Western) arrays using cell or tissue lysates
  • Purified protein arrays
  • Carbohydrate arrays
  • Lipids and other materials which can be arrayed on nitrocellulose
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)