Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z748056

Hanna carbon dioxide electrode filling solution

pkg of 4 × 30 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

packaging

pkg of 4 × 30 mL

manufacturer/tradename

Hanna HI 4005-40

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service