Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z747823

Whatman® Anotop® 25 syringe filters

sterile 0.2 μm, 50/pk

Synonym(s):

Whatman filter, syringe filter, syringe filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

material

Anopore membrane
alumina based membrane
alumina based membrane
polypropylene housing

sterility

sterile

packaging

pack of 50

manufacturer/tradename

Whatman 6809-2122
Whatman Article No. 28420397 (US reference)

parameter

100 mL sample volume

diam.

25 mm

pore size

0.2 μm

fitting

female Luer inlet (lock)
male Luer outlet

Looking for similar products? Visit Product Comparison Guide

Application

  • Cold sterilization of growth media
  • Phage and virus filtration
  • Removal of high molecular weight proteins or polymers
  • Liposome extrusion
  • Filtration of solvents for spectroanalysis and analytical sample preparation

Suitability

Suitable for filtration of aqueous or organic solutions.

Other Notes

Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.

The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.

Legal Information

Anotop is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service