Key Documents
Z744042
Whatman® PolyVENT and Sterivent intregal filters
PolyVENT 500, inlet 1/4 to 3/8 inch SB, outlet 1/4 to 3/8 inch SB
Synonym(s):
Whatman filter, disposable filter, filter Whatman
About This Item
UNSPSC Code:
41104900
material
PTFE membrane
polypropylene housing
sterility
non-sterile
manufacturer/tradename
Whatman 6713-1651
Whatman Article No. 28420174 (US reference)
packaging
1 ea of
diam.
16 mm
pore size
0.2 μm
fitting
inlet 1/4 to 3/8 inch SB
outlet 1/4 to 3/8 inch SB
General description
Whatman PolyVENT disc filters from Cytiva are integral venting filters that work bidirectionally to prevent contamination from entering containers like fermentation vessels during incubation.
Whatman PolyVENT venting filter
Draining or filling of incubators, fermentation tanks, and other vessels requires a venting filter capable of preventing bacterial contamination. With an integral PTFE filter membrane, Whatman PolyVENT acts as an industrial air filter media for sterilization* of gases entering bioreactors such as fermentation tanks.
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)