Z743904
ART® SoftFit L Reload System
Pure 300L, size 300 μL, reload insert, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
Select a Size
$795.00
Available to ship onApril 14, 2025Details
Select a Size
About This Item
reload insert
$795.00
Available to ship onApril 14, 2025Details
sterility
sterile
feature
Pure 300L
reload insert
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 3731-RI
technique(s)
PCR: suitable
size
300 μL
volume
300 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Reload towers offer an environmentally friendly packaging solution that effectively reduces plastic waste and minimizes required storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allow for quick and efficient identification of tip volume, streamlining your pipetting process.
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allows quick identification of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service