Z743901
ART® SoftFit L Reload System
Pure 20L, size 20 μL, hinged rack, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
Select a Size
$900.00
Available to ship onMay 02, 2025Details
Select a Size
About This Item
hinged rack
$900.00
Available to ship onMay 02, 2025Details
sterility
sterile
feature
Pure 20L
hinged rack
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 3721-HR
technique(s)
PCR: suitable
size
20 μL
volume
20 μL
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- The hinged rack of SoftFit-L™ pipette tips offer effortless one-hand access.
- Identify tip volume swiftly with color-coded inserts.
- Economical yet top-quality tips with a cylindrical collar and positive-stop design, minimizing attachment and ejection efforts.
- RNase, DNase, DNA, ATP, and endotoxin free.
- Each tip insert is marked with lot and product part numbers for easy traceability.
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allows quick identification of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service