Z743897
ART® SoftFit L Reload System
Pure 200L, size 1200 μL, hinged rack, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
About This Item
hinged rack
sterility
sterile
feature
Pure 200L
hinged rack
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 3751-HR
technique(s)
PCR: suitable
size
1200 μL
volume
1200 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allows quick identification of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service