Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743871

ART® Non-Filtered Tip

HLT YELLOW, size 200 μL, bulk racked, non-sterile

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 1000 EA
$712.00

$712.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
10 X 1000 EA
$712.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
200 μL
sterility:
non-sterile
feature:
HLT YELLOW
bulk racked
packaging:
pkg of 10X1000 ea
manufacturer/tradename:
Molecular BioProducts 3900

$712.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

sterility

non-sterile

feature

HLT YELLOW
bulk racked

packaging

pkg of 10X1000 ea

manufacturer/tradename

Molecular BioProducts 3900

technique(s)

PCR: suitable

size

200 μL

volume

200 μL

Looking for similar products? Visit Product Comparison Guide

General description

ART™ Non-filtered Hinged Rack Pipette Tips, which are made for the way you work, allows you to access tips with just one hand. You need not be concerned about your tips spilling on the bench or floor anymore. In just three simple steps, you can go from hinged to lift-off and maximize your productivity. Just unhinge, turn, and realign the cover to have a conventional lift-off when necessary. These tips are appropriate if contamination is not a concern.

Features and Benefits

  • Complete selection of non-filtered and specialty tips provides ART quality, performance, and reliability.
  • Racks allow saving time by offering quick and easy access to pipette tips.
  • The front latch closes and locks to provide safe and secure storage

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service