Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743831

ART® Non-Filtered Tip

ART 1250 REACH, size 1250 μL, hinged rack, non-sterile

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 768 EA
$488.00

$488.00


Available to ship onMay 05, 2025Details


Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
4 X 768 EA
$488.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1250 μL
sterility:
non-sterile
feature:
ART 1250 REACH
hinged rack
packaging:
pkg of 4X768 ea
manufacturer/tradename:
Molecular BioProducts 3152-HR

$488.00


Available to ship onMay 05, 2025Details


Notify Me

Get notified when this item is ready to ship via email.

sterility

non-sterile

feature

ART 1250 REACH
hinged rack

packaging

pkg of 4X768 ea

manufacturer/tradename

Molecular BioProducts 3152-HR

technique(s)

PCR: suitable

size

1250 μL

volume

1250 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

ART™ Non-filtered Hinged Rack Pipette Tips, which are made for the way you work, allows you to access tips with just one hand. You need not be concerned about your tips spilling on the bench or floor anymore. In just three simple steps, you can go from hinged to lift-off and maximize your productivity. Just unhinge, turn, and realign the cover to have a conventional lift-off when necessary. These tips are appropriate if contamination is not a concern.

Features and Benefits

  • Complete selection of non-filtered and specialty tips provides ART quality, performance, and reliability.
  • Racks allow saving time by offering quick and easy access to pipette tips.
  • The front latch closes and locks to provide safe and secure storage

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service